missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ NanoDrop™ 2000/2000c Spectrophotometers

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop One Microvolume UV-Vis Spectrophotometer.

Supplier:  Thermo Scientific™ ND2000CLAPTOP

 View more versions of this product

Catalog No. 13-400-412

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop One Microvolume UV-Vis Spectrophotometer.

The patented sample retention system of the NanoDrop 2000 and 2000c spectrophotometers allows a sample to be pipetted directly onto an optical measurement surface. Once the measurement is complete, the surfaces are simply wiped with a lint-free lab wipe. The NanoDrop 2000c integrates the sample retention system with cuvette capability.

NanoDrop 2000

  • Micro-volume sample size (0.5-2.0μL)
  • Easy to use—pipet sample directly onto the pedestal
  • No dilutions required, even for highly concentrated samples
  • Fast measurement time of less than 5 seconds
  • Full spectral output (190 - 840 nm)
  • Preconfigured methods for A260, A280, labeled DNA or Proteins, and much more
  • User-friendly software that includes custom methods and data export capabilities

NanoDrop 2000c

  • Combines all features of the NanoDrop 2000 with cuvette capability in one instrument
  • Advanced micro-volume pedestal technology
  • Dual modes—choose cuvette or pedestal
  • Broader concentration range for measuring very low or very high concentrations
  • Cuvette capability allows for kinetics (time or time/temperature studies) and cell culture (OD 600) measurements

NanoDrop 2000c - Cuvette

  • Heating: 37°C, ±0.5°C
  • Stirring: 150 - 850rpm
  • Z-Height: 8.5mm
  • Cuvette dimensions: 12.5 x 12.5mm, up to 48mmH
  • Cuvette path length options: 10, 5, 2 and 1mm
  • Type: Masked cuvette
  • Absorbance Range: 0.008 to 1.5
  • Measurement Time: <3 sec.
  • Weight: 2.1kg

Compatible with:

Professional versions of Microsoft™ Windows™ XP (32-bit) w/Service Pack (SP) 2 or later, Vista™ (32-bit), and Windows™ 7 (32 bit and 64 bit)

Specifications

Specifications

3% at (0.74 at 350 nm)
0.002
Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
USB
Microvolume Spectrophotometer, with cuvet capability
2 to 15,000 ng/μL (dsDNA) , 0.10 to 400mg/mL (BSA)
Xenon Flash
180 to 240 nm
12 VDC
14 x 20 cm
Laptop Computer
<5 sec.
37°; ±0.5°C
12 V DC
1.8 nm (FWHM at Hg 253.7)
0.5 to 2.0 μL
5 W
2.1 kg
Pedestal: 0 to 300 A Cuvette: 0 to 1.5 A
UL/CSA and CE
2 ng/μL to 15,000 ng/μL (dsDNA) ng/μL
20 cm
Pedestal: 2 ng/μL (dsDNA), 0.10 mg/mL (BSA) Cuvette: 0.4 ng/μL (dsDNA), 0.01 mg/mL (BSA)
2048-element Linear Silicon CCD Array
NanoDrop 2000c
UV-Vis Spectrophotometer with Laptop Computer
190 to 840 nm
Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
Laptop Computer
1
10, 5, 2, 1 mm
1000, 0.5 to 2.0 μL
Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
1 year parts and labor
±1 nm
14 cm
SDS
Documents

Documents

Promotions

Promotions

US Patents 6,628,382 and 6,809,826.