missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04171522

 View more versions of this product

Catalog No. AA4171522


Explore available promotions

Description

Description

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate is used as an anionic chromophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate
C10H10Na3O10S3
100 g
Soluble in water.
O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
455.33
434.31 (Anhydrous)
123409-01-8
MFCD00149263
1,3, 6,7-naphthalenetrisulfonic acid trisodium salt hydrate
BMCNZRJPDRPEDI-UHFFFAOYSA-N
naphthalene-1,3,6-trisulfonic acid;sodium;hydrate
123134558
Powder

Chemical Identifiers

123409-01-8
455.33
BMCNZRJPDRPEDI-UHFFFAOYSA-N
123134558
O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
C10H10Na3O10S3
MFCD00149263
1,3, 6,7-naphthalenetrisulfonic acid trisodium salt hydrate
naphthalene-1,3,6-trisulfonic acid;sodium;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-962-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only