missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate, Thermo Scientific Chemicals

$69.90 - $214.97

Chemical Identifiers

CAS 123409-01-8
Molecular Formula C10H10Na3O10S3
Molecular Weight (g/mol) 455.33
MDL Number MFCD00149263
InChI Key BMCNZRJPDRPEDI-UHFFFAOYSA-N
Synonym 1,3, 6,7-naphthalenetrisulfonic acid trisodium salt hydrate
PubChem CID 123134558
IUPAC Name naphthalene-1,3,6-trisulfonic acid;sodium;hydrate
SMILES O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4171522
View Documents
Thermo Scientific Chemicals
04171522
100 g N/A
 
AA4171536
View Documents
Thermo Scientific Chemicals
04171536
500 g N/A
 
Description

Description

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate is used as an anionic chromophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123409-01-8
455.33
BMCNZRJPDRPEDI-UHFFFAOYSA-N
123134558
O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
C10H10Na3O10S3
MFCD00149263
1,3, 6,7-naphthalenetrisulfonic acid trisodium salt hydrate
naphthalene-1,3,6-trisulfonic acid;sodium;hydrate

Specifications

123409-01-8
MFCD00149263
1,3, 6,7-naphthalenetrisulfonic acid trisodium salt hydrate
BMCNZRJPDRPEDI-UHFFFAOYSA-N
naphthalene-1,3,6-trisulfonic acid;sodium;hydrate
123134558
Powder
C10H10Na3O10S3
100 g
Soluble in water.
O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
455.33
434.31 (Anhydrous)
Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-962-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only