missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Naphthalene-1,4,5,8-tetracarboxylic Dianhydride 97.0+%, TCI America™
$206.97 - $1132.46
Chemical Identifiers
CAS | 81-30-1 |
---|---|
Molecular Formula | C14H4O6 |
Molecular Weight (g/mol) | 268.18 |
MDL Number | MFCD00006915 |
InChI Key | YTVNOVQHSGMMOV-UHFFFAOYSA-N |
Synonym | 1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride |
PubChem CID | 6678 |
SMILES | C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O |
Chemical Identifiers
81-30-1 | |
268.18 | |
YTVNOVQHSGMMOV-UHFFFAOYSA-N | |
6678 |
C14H4O6 | |
MFCD00006915 | |
1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride | |
C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O |
Specifications
81-30-1 | |
MFCD00006915 | |
1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride | |
C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O | |
6678 | |
≥97.0% (HPLC) | |
Naphthalene-1,4,5,8-tetracarboxylic Dianhydride |
C14H4O6 | |
25 g | |
YTVNOVQHSGMMOV-UHFFFAOYSA-N | |
268.18 | |
268.18 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : QK3695000
TSCA : Yes