missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neodymium(III) isopropoxide, 99% (REO), 2.5% w/v in toluene/isopropanol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 036573AE

 View more versions of this product

Catalog No. AA36573AE


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Neodymium(III) isopropoxide
Blue
C9H24NdO3
MFCD00058819,MFCD00058819
UN1993
neodymium i-propoxide, neodymium iii isopropoxide
[Nd].CC(C)O.CC(C)O.CC(C)O
324.53
321.3
2.5% w/v in toluene/isopropanol
19236-15-8
(REO)
Nd[OCH(CH3)2]3
100 mL
Moisture sensitive
WOCLHTSADRFWMF-UHFFFAOYSA-N
neodymium;propan-2-ol
25199945
99%
Liquid

Chemical Identifiers

19236-15-8
324.53
WOCLHTSADRFWMF-UHFFFAOYSA-N
25199945
C9H24NdO3
MFCD00058819,MFCD00058819
neodymium i-propoxide, neodymium iii isopropoxide
[Nd].CC(C)O.CC(C)O.CC(C)O
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H318-H336-H315
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
May cause drowsiness or dizziness.
Causes skin irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336-H361d-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only