Learn More
Neodymium(III) isopropoxide, 99% (REO), 2.5% w/v in toluene/isopropanol, Thermo Scientific Chemicals
$222.33 - $617.86
Chemical Identifiers
CAS | 19236-15-8 |
---|---|
Molecular Formula | C9H24NdO3 |
Molecular Weight (g/mol) | 324.53 |
MDL Number | MFCD00058819,MFCD00058819 |
InChI Key | WOCLHTSADRFWMF-UHFFFAOYSA-N |
Synonym | neodymium i-propoxide, neodymium iii isopropoxide |
PubChem CID | 25199945 |
SMILES | [Nd].CC(C)O.CC(C)O.CC(C)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19236-15-8 | |
324.53 | |
WOCLHTSADRFWMF-UHFFFAOYSA-N | |
25199945 |
C9H24NdO3 | |
MFCD00058819,MFCD00058819 | |
neodymium i-propoxide, neodymium iii isopropoxide | |
[Nd].CC(C)O.CC(C)O.CC(C)O |
Specifications
19236-15-8 | |
C9H24NdO3 | |
MFCD00058819,MFCD00058819 | |
UN1993 | |
neodymium i-propoxide, neodymium iii isopropoxide | |
[Nd].CC(C)O.CC(C)O.CC(C)O | |
324.53 | |
321.3 | |
2.5% w/v in toluene/isopropanol | |
Neodymium(III) isopropoxide |
Blue | |
Nd[OCH(CH3)2]3 | |
25 mL | |
Moisture sensitive | |
WOCLHTSADRFWMF-UHFFFAOYSA-N | |
tris(propan-2-ol) neodymium | |
25199945 | |
99% | |
Liquid | |
(REO) |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H318-H336-H315
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
May cause drowsiness or dizziness.
Causes skin irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H361d-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only