missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Neon™ Transfection System 10 μL Kit
Neon™ Transfection System 10 μL Kit
Supplier: Invitrogen™ MPK1025
Description
The Neon™ Transfection System 10 μL Kit is designed specifically for use with the MPK5000. Use this kit for transfection volumes of 10 μL, containing 5 × 104–2 × 105 adherent cells or 1 × 105–5 × 105 suspension cells. Cells that have been transfected using the included 10μL Neon™ Tips are ready to be washed and plated into a multi-well culture dish.Specifications
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum | |
Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA, Synthetic siRNA | |
Neon™ Transfection System | |
25 x 2 | |
Transfection System | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type | |
10 μl |
The Neon™ Transfection System 10μL Kit comes with 1 mL resuspension buffer R, 1 mL resuspension buffer T, and 75 mL E electrolytic buffer in one box and 25 reaction delivery tips and 5 electroporation tubes in a separate box. Refer to the Neon™ Transfection System users manual for protocol instructions. Store the buffer box 4°C and the tips & tubes box at room temperature. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Electroporation | |
Transfection | |
Neon™ | |
25 x 2 Reactions | |
Room Temperature | |
Neon™ Transfection System |
For Research Use Only. Not for use in diagnostic procedures.