missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Neon™ Transfection System 10 μL Kit
Neon™ Transfection System 10 μL Kit
$1060.00 - $3105.00
Specifications
Classification | Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum |
---|---|
Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Sample Type | Plasmid DNA, Synthetic siRNA |
Transfection Technique | Electroporation |
Description
The Neon™ Transfection System 10 μL Kit is designed specifically for use with the MPK5000. Use this kit for transfection volumes of 10 μL, containing 5 × 104–2 × 105 adherent cells or 1 × 105–5 × 105 suspension cells. Cells that have been transfected using the included 10μL Neon™ Tips are ready to be washed and plated into a multi-well culture dish.Specifications
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Electroporation | |
Transfection | |
Transfection System | |
Room Temperature | |
Neon™ Transfection System |
Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA, Synthetic siRNA | |
Neon™ Transfection System | |
Neon™ | |
Yes | |
2x10^4 - 6 x10^8 cells; Varies by cell type |
For Research Use Only. Not for use in diagnostic procedures.