Invitrogen™ Neon™ Transfection System 10 μL Kit DFS Item
Neon™ Transfection System 10 μL Kit
Supplier:  Invitrogen™ MPK1096
 View more versions of this product
Catalog No. MPK1096
Explore available promotions
Includes: 3 x 1mL Resuspension Buffer R, 3 x 1mL Resuspension Buffer T, 2 x 150mL Electrolytic Buffer E, 96 x 10μL Neon tips, 20 Neon electroporation tubes
Description
Description
  • Animal origin-free, serum-free and designed specifically for use with the Neon Transfection System
  • Used for transfection volumes of 10μL, containing 5 x 104-2 x 105 adherent cells or 1 x 106-5 x 105 suspension cells
  • Cells that have been transfected using included 10μL Neon Tips are ready to be washed and plated into multiwell culture dish
Adenoviral shRNA, Cell Culture, Lentiviral shRNA & miRNA, Plasmid Transfection, RNAi, RNAi Transfection, RNAi, Epigenetics & Non-Coding RNA Research, Retroviral RNAi, Stem Cell & Primary Cell Transfections, Synthetic siRNA Transfection, Transfection, Vector-Based RNAi, siRNA
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum
Stem Cells, Primary Cells, Hard-to-Transfect Cells
Plasmid DNA, Synthetic siRNA
10 x 10^4 - 2 x 10^5 cells/mL (Adherent Cells), 1 x 10^6 - 5 x 10^5 cells/mL
Transfection
Neon™
96 x 2 Reactions
Room Temperature
Neon™ Transfection System
• 3 x 1 mL Resuspension Buffer R
• 3 x 1 mL Resuspension Buffer T
• 2 x 150 mL Electrolytic Buffer E
• 96 x 10 μl Neon™ tips
• 20 Neon™ electroporation tubes
Store the buffer box 4°C and the tips and tubes box at room temperature.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Electroporation
Neon™ Transfection System
96 x 2
Transfection System
Yes
2x10^4 - 6 x10^8 cells; Varies by cell type
10μl, 10 μl
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.