missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nerol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 376540250

 View more versions of this product

Catalog No. AC376540250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nerol
96.0
Colorless
225°C to 227°C
Authentic
Glass bottle
1.4720 to 1.479
MFCD00063204
01, IV, 2276
15, 6560
Solubility in water: practically insoluble in water. Other solubilities: soluble in absolute alcohol
CC(C)=CCC\C(C)=C/CO
154.25
CHEBI:29452
97%
106-25-2
100.0
0.8750g/mL
100°C
96% min. (GC)
C10H18O
(CH3)2CCH(CH2)2C(CH3)CHCH2OH
25 mL
0.875
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
GLZPCOQZEFWAFX-YFHOEESVSA-N
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
643820
154.25
Liquid

Chemical Identifiers

106-25-2
154.25
GLZPCOQZEFWAFX-YFHOEESVSA-N
643820
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
C10H18O
MFCD00063204
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
CHEBI:29452
CC(C)=CCC\C(C)=C/CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-378-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only