missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nerol, 97%, Thermo Scientific Chemicals

$55.22 - $109.17

Chemical Identifiers

CAS 106-25-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00063204
InChI Key GLZPCOQZEFWAFX-YFHOEESVSA-N
Synonym nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
PubChem CID 643820
ChEBI CHEBI:29452
IUPAC Name (2Z)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(C)=CCC\C(C)=C/CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC376540250
SDS View Documents
Thermo Scientific Chemicals
376540250
25 mL Glass bottle
Each for $55.22
 
AC376541000
SDS View Documents
Thermo Scientific Chemicals
376541000
100 mL Glass bottle
Each for $109.17
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-25-2
154.25
GLZPCOQZEFWAFX-YFHOEESVSA-N
643820
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
C10H18O
MFCD00063204
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
CHEBI:29452
CC(C)=CCC\C(C)=C/CO

Specifications

106-25-2
100.0
0.8750g/mL
100°C
96% min. (GC)
C10H18O
(CH3)2CCH(CH2)2C(CH3)CHCH2OH
25 mL
0.875
nerol, cis-geraniol, neryl alcohol, z-3,7-dimethylocta-2,6-dien-1-ol, z-geraniol, 2z-3,7-dimethylocta-2,6-dien-1-ol, 2,6-octadien-1-ol, 3,7-dimethyl-, 2z, nerol natural, cis-3,7-dimethyl-2,6-octadien-1-ol, z-3,7-dimethyl-2,6-octadien-1-ol
GLZPCOQZEFWAFX-YFHOEESVSA-N
(2Z)-3,7-dimethylocta-2,6-dien-1-ol
643820
154.25
Liquid
96.0
Colorless
225°C to 227°C
Authentic
Glass bottle
1.4720 to 1.479
MFCD00063204
01, IV, 2276
15, 6560
Solubility in water: practically insoluble in water. Other solubilities: soluble in absolute alcohol
CC(C)=CCC\C(C)=C/CO
154.25
CHEBI:29452
97%
Nerol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-378-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only