Neutec Group Spin Air Microbial Air Samplers
Used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications
Supplier:  Neutec Group 90005502
 View more versions of this product
Catalog No. 14-910-438
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Microbial air sampling slave unit must be connected to a main Spin Air unit. When connected to a main unit it allows for duplicate sampling.
  • Rotating plate holder for even surface spread of micro organisms (Except the Basic Air model)
  • Sampling by volume (not timed) for higher accuracy and reproducibility
  • Utilizes standard 60mm or 90mm agar plates which are easy to find and inexpensive
  • Rechargeable battery operated for remote sampling
  • Delayed activation mode to avoid collector’s contamination contribution
  • Optional duplicate simultaneous sampling with low cost daughter unit (Spin Air model only)
  • Bar code connectivity for faster easier collection of sample, location and user information (Spin Air Model)
  • Wide range of adaptors and accessories plastic and aluminum choices in 60mm, 90mm and Rodac sizes
Specifications
Specifications
Spin Air Microbial Air Sampler Daughter Unit