missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neutec Group Spin Air Microbial Air Samplers
Used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications
$13393.47 - $17913.50
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
14-910-435
|
Neutec Group
90005500 |
Spin Air Microbial Air Sampler |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14-910-438
|
Neutec Group
90005502 |
Spin Air Microbial Air Sampler Daughter Unit |
Each for $13,393.47
|
|
|||||
Description
Microbial air sampling slave unit must be connected to a main Spin Air unit. When connected to a main unit it allows for duplicate sampling.- Rotating plate holder for even surface spread of micro organisms (Except the Basic Air model)
- Sampling by volume (not timed) for higher accuracy and reproducibility
- Utilizes standard 60mm or 90mm agar plates which are easy to find and inexpensive
- Rechargeable battery operated for remote sampling
- Delayed activation mode to avoid collector’s contamination contribution
- Optional duplicate simultaneous sampling with low cost daughter unit (Spin Air model only)
- Bar code connectivity for faster easier collection of sample, location and user information (Spin Air Model)
- Wide range of adaptors and accessories plastic and aluminum choices in 60mm, 90mm and Rodac sizes