missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ New Brunswick Innova™ 42/42 R - Stackable Incubator Shaker

Versatile and reliable. The Eppendorf Innova 42 and 42R (refrigerated) incubator shakers are stackable, can be placed on or under the bench, and come with a static shelf to be used to incubate agar plates while shaking cultures in flasks (Optional: static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan.

Supplier:  Eppendorf™ M13350004

 View more versions of this product

Catalog No. 05400157

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • High capacity in a small footprint: for example, 4 x 5 L or 2 x 6 L flasks on universal platform; device footprint 4731 cm2 /733 in.2 (63.5 cm/25 in. width, 74.5 cm/29.3 in. depth)
  • Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes, and can be reached easily even in a double stack
  • Easy to clean: Spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
  • Lab position flexibility: Lab position on or under bench, 2x stackable
  • Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
  • Sample evaporation protection: Bottom spill pan usable as water reservoir; optional culture drawer, for Agar plates, for example
  • Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
  • Reliable 24/7 with high loads: Proven and tested Innova triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
  • Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
  • Options and Accessories: Orbit 19 or 25 mm, photosynthetic light bank, UV germicidal light, culture drawer for evaporation protection of Agar plates, additional static shelves (one included), humidity monitoring, remote monitoring with VisioNize™ Lab Suite and more
  • Speed: 25 to 400 rpm, 25 to 300 rpm when using flasks >500 mL in stacked devices
  • Certifications/Compliance: 2014/35/EU: EN 61010-1, EN 61010-2-010, EN 61010-2-051, 2014/30/EU: EN 61326-1, EN 55011 (Class A), 2011/65/EU: EN IEC 63000, IEC 61010-1, IEC 61010-2-010, IEC 61010-2-051, UL 61010-1, UL 61010-2-010, UL 61010-2-051, CAN/CSA C22.2 No. 61010-1-12, CSA C22.2 No. 61010-2-010, CSA C22.2 No. 61010-2-051, IEC 61326-1, CISPR 11, FCC 47 CFR Part 15 (Class A)
Specifications

Specifications

19 mm
25 to 400 rpm
120 V
6 L Flasks
20 to 35 lb.
±0.25°C at 37°C
Aluminum Platform
46 x 46 cm (18 x 18 in.)
32.2 in.
81.8 cm
25 in.
63.5 cm
266 lb.
Innova 42R
Plasmid DNA production and protein expression, Biofuel research, Grow bacteria, yeast, fungus, photosynthetic organisms
Accessories
Universal/Dedicated
0.01 sec. to 95 hr. 59 min., Continuous
Yes
9 to 15.8 kg
120 V 50/60 Hz
20°C below ambient to 80°C
Incubator Shaker
68°F below ambient to 176°F
60 Hz
51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.)
63.5 x 74.5 x 81.8 cm / 25 x 29.3 x 32.8 in.
18.8 in.
47.7 cm
20.4 in.
51.7 cm
121 kg
LED Digital Electric Display
20 to 80%, Non-condensing
USB, Ethernet
Universal/Dedicated
3 Year
Videos
SDS
Documents

Documents

Promotions

Promotions