missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ New Brunswick Innova™ 42/42 R - Stackable Incubator Shaker
Versatile and reliable. The Eppendorf Innova 42 and 42R (refrigerated) incubator shakers are stackable, can be placed on or under the bench, and come with a static shelf to be used to incubate agar plates while shaking cultures in flasks (Optional: static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan.
$21507.58 - $24429.71
Specifications
Product Type | Incubator Shaker |
---|---|
Dimensions (LxW) Platform | 46 x 46 cm (18 x 18 in.) |
Catalog Number | Mfr. No. | Model | Refrigerated | Orbit | Electrical Requirements | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Refrigerated | Orbit | Electrical Requirements | Price | Quantity | ||
05400155
|
Eppendorf™
M13350000 |
Innova 42 | No | 19 mm | 120 V 50/60 Hz |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
05400157
|
Eppendorf™
M13350004 |
Innova 42R | Yes | 19 mm | 120 V 50/60 Hz |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
05400159
|
Eppendorf™
M13350010 |
Innova 42 | No | 25 mm | 120 V 50/60 Hz |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
05400162
|
Eppendorf™
M13350014 |
Innova 42R | Yes | 25 mm | 120 V 50/60 Hz |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- High capacity in a small footprint: for example, 4 x 5 L or 2 x 6 L flasks on universal platform; device footprint 4731 cm2 /733 in.2 (63.5 cm/25 in. width, 74.5 cm/29.3 in. depth)
- Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes, and can be reached easily even in a double stack
- Easy to clean: Spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
- Lab position flexibility: Lab position on or under bench, 2x stackable
- Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
- Sample evaporation protection: Bottom spill pan usable as water reservoir; optional culture drawer, for Agar plates, for example
- Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
- Reliable 24/7 with high loads: Proven and tested Innova triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
- Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
- Options and Accessories: Orbit 19 or 25 mm, photosynthetic light bank, UV germicidal light, culture drawer for evaporation protection of Agar plates, additional static shelves (one included), humidity monitoring, remote monitoring with VisioNize™ Lab Suite and more
- Speed: 25 to 400 rpm, 25 to 300 rpm when using flasks >500 mL in stacked devices
- Certifications/Compliance: 2014/35/EU: EN 61010-1, EN 61010-2-010, EN 61010-2-051, 2014/30/EU: EN 61326-1, EN 55011 (Class A), 2011/65/EU: EN IEC 63000, IEC 61010-1, IEC 61010-2-010, IEC 61010-2-051, UL 61010-1, UL 61010-2-010, UL 61010-2-051, CAN/CSA C22.2 No. 61010-1-12, CSA C22.2 No. 61010-2-010, CSA C22.2 No. 61010-2-051, IEC 61326-1, CISPR 11, FCC 47 CFR Part 15 (Class A)
Specifications
Incubator Shaker | |
46 x 46 cm (18 x 18 in.) |