Fisherbrand™ Newport Industrial Polyurethane Chair - Medium Bench Height with Chrome Foot Ring and Casters in Polyurethane
Fisherbrand™ Newport Polyurethane Chairs are height for 36 in. work surfaces and feature ergonomic adjustments, handle in the back rest, chrome foot ring, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
 View more versions of this product
Catalog No. 15000574
Explore available promotions
Description
Description
  • Adjustable from 19.75-27.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floor
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Pneumatic
No
Yes
15.42 kg
Yes
136 kg
No
Gray
Medium Bench Height
15 year parts, lifetime on pneumatic lift
Yes
19.75 to 27.5 in.
No
Black dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
34 lb.
Nylon Reinforced Fiberglass
300 lb.
Polyurethane