Learn More
Nextteq™ Gastec™ Detector Tubes: Hydrogen Sulfide Gas
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Supplier: Nextteq™ 4LT
Description
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
| Hydrogen Sulfide | |
| 0.5 to 2 | |
| 30 sec. to 2 min. | |
| 0°C to 40°C |
| 0.1 to 4ppm | |
| 10 | |
| 32°F to 104°F |