Learn More
Nextteq™ Gastec™ Detector Tubes: Hydrogen Sulfide Gas
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
$292.06 - $305.47
Specifications
Components Measured | Hydrogen Sulfide |
---|---|
Quantity | 10 |
Temperature Range (English) | 32°F to 104°F |
Temperature Range (Metric) | 0°C to 40°C |
Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | Price | Quantity | |||
19-026-286
|
Nextteq™
4H |
Hydrogen Sulfide | 10 to 4000ppm | 30 sec. to 7.5 min. |
Pack of 10 Each for $297.38
|
|
|||
19-026-290
|
Nextteq™
4HT |
Hydrogen Sulfide | 1 to 40% | 45 sec. to 3 min. |
Pack of 10 Each for $305.47
|
|
|||
19-026-288
|
Nextteq™
4HM |
Hydrogen Sulfide | 25 to 1600ppm | 30 sec. to 1.5 min. |
Pack of 10 Each for $297.38
|
|
|||
19-026-295
|
Nextteq™
4M |
Hydrogen Sulfide | 12.5 to 500ppm | 30 sec. to 1.5 min. |
Pack of 10 Each for $297.38
|
|
|||
19026294
|
Nextteq™
4LT |
Hydrogen Sulfide | 0.1 to 4ppm | 30 sec. to 2 min. |
Pack of 10 Each for $292.06
|
|
|||
19-026-287
|
Nextteq™
4HH |
Hydrogen Sulfide | 0.1 to 4% | 45 to 30 sec. |
Pack of 10 Each for $297.38
|
|
|||
19-026-291
|
Nextteq™
4L |
Hydrogen Sulfide | 1 to 240ppm | 30 sec. to 7.5 min. |
Pack of 10 Each for $297.38
|
|
|||
19-026-271
|
Nextteq™
4LB |
Hydrogen Sulfide | 0.5 to 12ppm | 30 sec. to 1.5 min. |
Pack of 10 Each for $297.38
|
|
|||
Description
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Hydrogen Sulfide | |
10 | |
32°F to 104°F | |
0°C to 40°C |