Nextteq™ Gastec™ Detector Tubes: Hydrogen Sulfide Gas
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Supplier:  Nextteq™ 4M
 View more versions of this product
Catalog No. 19-026-295
Explore available promotions
Description
Description
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Specifications
Hydrogen Sulfide
0.5 to 2
30 sec. to 1.5 min.
0°C to 40°C
12.5 to 500ppm
10
32°F to 104°F