missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Niacin, Powder, USP, 98-102%, Spectrum™ Chemical
NI100, 59-67-6, C6H5NO2
Supplier: Spectrum Chemical Mfg Cor NI100500GM
Description
Spectrum™ Chemical Niacin, Powder, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
59-67-6 | |
0.001 | |
Poly Bottle | |
MFCD00006391 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
pyridine-3-carboxylic acid | |
98 to 102% |
1 | |
0.01 | |
C6H5NO2 | |
500 g | |
OC(=O)C1=CC=CN=C1 | |
123.11 | |
USP |
Chemical Identifiers
59-67-6 | |
123.11 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
OC(=O)C1=CC=CN=C1 |
C6H5NO2 | |
MFCD00006391 | |
pyridine-3-carboxylic acid |