missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Niacin, Powder, USP, 98-102%, Spectrum™ Chemical
NI100, 59-67-6, C6H5NO2
$528.31 - $10634.83
Chemical Identifiers
CAS | 59-67-6 |
---|---|
Molecular Formula | C6H5NO2 |
Molecular Weight (g/mol) | 123.11 |
MDL Number | MFCD00006391 |
InChI Key | PVNIIMVLHYAWGP-UHFFFAOYSA-N |
IUPAC Name | pyridine-3-carboxylic acid |
SMILES | OC(=O)C1=CC=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18604781
|
Spectrum Chemical Mfg Cor
NI100500GM |
500 g | Poly Bottle |
Each for $528.31
|
|
||||
18604783
|
Spectrum Chemical Mfg Cor
NI10012KGBL |
12 kg | Poly Pail |
Each for $3,217.60
|
|
||||
18604784
|
Spectrum Chemical Mfg Cor
NI10025KGBL |
25 kg | Fiber Drum |
Each for $6,686.91
|
|
||||
18604785
|
Spectrum Chemical Mfg Cor
NI10050KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Niacin, Powder, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
59-67-6 | |
123.11 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
OC(=O)C1=CC=CN=C1 |
C6H5NO2 | |
MFCD00006391 | |
pyridine-3-carboxylic acid |
Specifications
59-67-6 | |
0.001 | |
Poly Bottle | |
MFCD00006391 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
pyridine-3-carboxylic acid | |
98 to 102% |
1 | |
0.01 | |
C6H5NO2 | |
500 g | |
OC(=O)C1=CC=CN=C1 | |
123.11 | |
USP |