Niacin, Powder, USP, 98-102%, Spectrum™ Chemical
NI100, 59-67-6, C6H5NO2
Supplier:  Spectrum Chemical Mfg Cor NI10012KGBL
 View more versions of this product
Catalog No. 18604783
Explore available promotions
Description
Description
Spectrum™ Chemical Niacin, Powder, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
59-67-6
0.001
Poly Pail
MFCD00006391
PVNIIMVLHYAWGP-UHFFFAOYSA-N
pyridine-3-carboxylic acid
98 to 102%
1
0.01
C6H5NO2
12 kg
OC(=O)C1=CC=CN=C1
123.11
USP
Chemical Identifiers
59-67-6
123.11
PVNIIMVLHYAWGP-UHFFFAOYSA-N
OC(=O)C1=CC=CN=C1
C6H5NO2
MFCD00006391
pyridine-3-carboxylic acid