missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel acetylacetonate, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 128260050

 View more versions of this product

Catalog No. AC128260050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel acetylacetonate
95.0
230°C
220°C (11.0 mmHg)
95% min. (Complexometry)
C10H14NiO4
5 g
15, 6585
Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform and benzene,insoluble in ether and ligroin
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
256.91
256.9
Liquid
3264-82-2
100.0
Yellow
Authentic
Glass bottle
MFCD00000024
05,471; 06,417; 07,250; 08,42; 17,201
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
BMGNSKKZFQMGDH-FDGPNNRMSA-L
nickel(2+);(E)-4-oxopent-2-en-2-olate
53384569
96%

Chemical Identifiers

3264-82-2
256.91
BMGNSKKZFQMGDH-FDGPNNRMSA-L
53384569
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
C10H14NiO4
MFCD00000024
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
nickel(2+);(E)-4-oxopent-2-en-2-olate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: I

GHS Signal Word: Danger

EINECSNumber : 221-875-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only