missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel acetylacetonate, 96%, Thermo Scientific Chemicals

$73.71 - $677.16

Chemical Identifiers

CAS 3264-82-2
Molecular Formula C10H14NiO4
Molecular Weight (g/mol) 256.91
MDL Number MFCD00000024
InChI Key BMGNSKKZFQMGDH-FDGPNNRMSA-L
Synonym nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
PubChem CID 53384569
IUPAC Name nickel(2+);(E)-4-oxopent-2-en-2-olate
SMILES [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128260050
SDS View Documents
Thermo Scientific Chemicals
128260050
5 g Glass bottle
Each for $73.71
 
AC128260250
SDS View Documents
Thermo Scientific Chemicals
128260250
25 g Glass bottle
Each for $228.54
 
AC128261000
SDS View Documents
Thermo Scientific Chemicals
128261000
100 g Glass bottle
Each for $677.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3264-82-2
256.91
BMGNSKKZFQMGDH-FDGPNNRMSA-L
53384569
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
C10H14NiO4
MFCD00000024
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
nickel(2+);(E)-4-oxopent-2-en-2-olate

Specifications

3264-82-2
100.0
Yellow
Authentic
Glass bottle
MFCD00000024
05,471; 06,417; 07,250; 08,42; 17,201
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
BMGNSKKZFQMGDH-FDGPNNRMSA-L
nickel(2+);(E)-4-oxopent-2-en-2-olate
53384569
96%
Nickel acetylacetonate
95.0
230°C
220°C (11.0 mmHg)
95% min. (Complexometry)
C10H14NiO4
5 g
15, 6585
Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform and benzene,insoluble in ether and ligroin
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
256.91
256.9
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: I

GHS Signal Word: Danger

EINECSNumber : 221-875-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only