Learn More
Nickel acetylacetonate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 128261000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel acetylacetonate | |
95.0 | |
230°C | |
220°C (11.0 mmHg) | |
95% min. (Complexometry) | |
C10H14NiO4 | |
100 g | |
15, 6585 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform and benzene,insoluble in ether and ligroin | |
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
256.91 | |
256.9 | |
Liquid |
3264-82-2 | |
100.0 | |
Yellow | |
Authentic | |
Glass bottle | |
MFCD00000024 | |
05,471; 06,417; 07,250; 08,42; 17,201 | |
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii | |
BMGNSKKZFQMGDH-FDGPNNRMSA-L | |
nickel(2+);(E)-4-oxopent-2-en-2-olate | |
53384569 | |
96% |
Chemical Identifiers
3264-82-2 | |
256.91 | |
BMGNSKKZFQMGDH-FDGPNNRMSA-L | |
53384569 | |
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
C10H14NiO4 | |
MFCD00000024 | |
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii | |
nickel(2+);(E)-4-oxopent-2-en-2-olate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: I
GHS Signal Word: Danger
EINECSNumber : 221-875-7
RUO – Research Use Only