missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) acetate tetrahydrate, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 415541000

 View more versions of this product

Catalog No. AC415541000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel(II) acetate tetrahydrate
6018-89-9,373-02-4
100.0
Orange
98.5% min
C4H14NiO8
MFCD00066973
14,288; 16,308
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
OINIXPNQKAZCRL-UHFFFAOYSA-L
nickel(2+);diacetate;tetrahydrate
62601
99%
Powder
Extra Pure
98.5
250.0°C
Authentic
Plastic bottle
(CH3CO2)2Ni·4H2O
100 g
14, 6498
Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
248.84
248.86
Extra Pure

Chemical Identifiers

6018-89-9
248.84
OINIXPNQKAZCRL-UHFFFAOYSA-L
62601
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
C4H14NiO8
MFCD00066973
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
nickel(2+);diacetate;tetrahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only