Learn More
Nickel(II) acetate tetrahydrate, 99%, extra pure, Thermo Scientific Chemicals
$60.10 - $137.39
Chemical Identifiers
CAS | 6018-89-9 |
---|---|
Molecular Formula | C4H14NiO8 |
Molecular Weight (g/mol) | 248.84 |
MDL Number | MFCD00066973 |
InChI Key | OINIXPNQKAZCRL-UHFFFAOYSA-L |
Synonym | nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate |
PubChem CID | 62601 |
IUPAC Name | nickel(2+);diacetate;tetrahydrate |
SMILES | O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC415541000
|
Thermo Scientific Chemicals
415541000 |
100 g | Plastic bottle |
Each for $60.10
|
|
||||
AC415545000
|
Thermo Scientific Chemicals
415545000 |
500 g | Plastic bottle |
Each for $137.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6018-89-9 | |
248.84 | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
62601 | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
C4H14NiO8 | |
MFCD00066973 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
nickel(2+);diacetate;tetrahydrate |
Specifications
6018-89-9,373-02-4 | |
100.0 | |
Orange | |
98.5% min | |
C4H14NiO8 | |
MFCD00066973 | |
14,288; 16,308 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
nickel(2+);diacetate;tetrahydrate | |
62601 | |
99% | |
Powder |
98.5 | |
250.0°C | |
Authentic | |
Plastic bottle | |
(CH3CO2)2Ni·4H2O | |
100 g | |
14, 6498 | |
Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O | |
248.84 | |
248.86 | |
Extra Pure | |
Nickel(II) acetate tetrahydrate, Extra Pure |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
GHS Signal Word: Danger
RUO – Research Use Only