missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) bromide, 99%

CAS: 13462-88-9 | Br2Ni | 218.53 g/mol

Supplier:  Thermo Scientific Chemicals 203360500

 View more versions of this product

Catalog No. AC203360500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel(II) bromide
13462-88-9
98.5
Crystalline Powder and/or Chunks
98.5% min. (on dry substance) (Argentometry)
NiBr2
04,351
nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
IPLJNQFXJUCRNH-UHFFFAOYSA-L
dibromonickel
278492
99%
963.0°C
Brown
100.0
50 g
Br2Ni
MFCD00011141
15, 6587
Solubility in water: 567g/L (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether
[Ni](Br)Br
218.53
218.53
Glass bottle

Chemical Identifiers

13462-88-9
218.53
IPLJNQFXJUCRNH-UHFFFAOYSA-L
278492
[Ni](Br)Br
Br2Ni
MFCD00011141
nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
dibromonickel
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
May cause allergy or asthma symptoms or brea

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
Avoid release to the environment.
Obtain

GHS Signal Word: Danger

EINECSNumber : 236-665-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only