missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) bromide hydrate, 98%, for analysis, Thermo Scientific Chemicals

$189.10 - $189.10

Chemical Identifiers

CAS 207569-11-7
Molecular Formula Br2Ni
Molecular Weight (g/mol) 218.50
MDL Number MFCD00149806
InChI Key IPLJNQFXJUCRNH-UHFFFAOYSA-L
Synonym nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci
PubChem CID 57377118
SMILES [Ni++].[Br-].[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC214871000
SDS View Documents
Thermo Scientific Chemicals
214871000
100 g Glass bottle
Each for $189.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207569-11-7
218.50
IPLJNQFXJUCRNH-UHFFFAOYSA-L
57377118
Br2Ni
MFCD00149806
nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci
[Ni++].[Br-].[Br-]

Specifications

963.0°C
Orange
100.0
Crystalline Powder or Needles
97.5% min. (on dry substance) (Argentometry)
NiBr2·xH2O
04,351
nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci
IPLJNQFXJUCRNH-UHFFFAOYSA-L
dibromonickel;hydrate
57377118
98%
Glass bottle
207569-11-7,13462-88-9
97.5
5 to 7 (5% aq. soln.)
100 g
Br2Ni
MFCD00149806
10, 6340
Solubility in water: soluble in water. Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether
[Ni++].[Br-].[Br-]
218.50
218.53
Analytical
Nickel(II) bromide hydrate, 98%
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
M

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain speci

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only