missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1554022

 View more versions of this product

Catalog No. AAA1554022

Explore available promotions

Description

Description

Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

Solubility
Very soluble in water. Soluble in ethanol.

Notes
As an oxidizer, it is incompatible with reducing agents.
Specifications

Specifications

137°C
56.7°C
100 g
Ni(NO3)2·6H2O
UN2725
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
AOPCKOPZYFFEDA-UHFFFAOYSA-N
nickel(2+);dinitrate;hexahydrate
61630
98%
Hygroscopic
Nickel(II) nitrate hexahydrate
13478-00-7
H12N2NiO12
MFCD00149805
14,6513
Very soluble in water. Soluble in ethanol.
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
290.79
290.80 (182.70 Anhydrous)
Odorless
2.05

Chemical Identifiers

13478-00-7
290.79
AOPCKOPZYFFEDA-UHFFFAOYSA-N
61630
H12N2NiO12
MFCD00149805
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H350-H360-H372-H341-H318-H302-H332-H315-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P362-P370+P378q-P501c

H272-H302+H332-H315-H317-H318-H334-H335-H341-H350i-H360D-H372

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NICKEL NITRATE

EINECSNumber : 236-068-5

RTECSNumber : QR7300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only