Learn More
Nickel(II) nitrate hexahydrate, 98%, Thermo Scientific Chemicals
$58.99 - $858.30
Chemical Identifiers
CAS | 13478-00-7 |
---|---|
Molecular Formula | H12N2NiO12 |
Molecular Weight (g/mol) | 290.79 |
MDL Number | MFCD00149805 |
InChI Key | AOPCKOPZYFFEDA-UHFFFAOYSA-N |
Synonym | nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 |
PubChem CID | 61630 |
SMILES | O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1554022
|
Thermo Scientific Chemicals
A1554022 |
100 g |
Each for $58.99
|
|
||||||
AAA1554036
|
Thermo Scientific Chemicals
A1554036 |
500 g |
Each for $115.59
|
|
||||||
AAA155400B
|
Thermo Scientific Chemicals
A155400B |
1000 g |
Each for $193.58
|
|
||||||
AAA155400I
|
Thermo Scientific Chemicals
A155400I |
5000 g |
Each for $858.30
|
|
||||||
AAA15540A4
|
Thermo Scientific Chemicals
A15540A4 |
2.5 kg |
Each for $434.63
|
|
||||||
Description
Nickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel(II) nitrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.
Solubility
Very soluble in water. Soluble in ethanol.
Notes
As an oxidizer, it is incompatible with reducing agents.
Chemical Identifiers
13478-00-7 | |
290.79 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
61630 |
H12N2NiO12 | |
MFCD00149805 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
137°C | |
13478-00-7 | |
H12N2NiO12 | |
MFCD00149805 | |
14,6513 | |
Very soluble in water. Soluble in ethanol. | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
290.79 | |
290.80 (182.70 Anhydrous) | |
Odorless | |
2.05 |
56.7°C | |
100 g | |
Ni(NO3)2·6H2O | |
UN2725 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
nickel(2+);dinitrate;hexahydrate | |
61630 | |
98% | |
Hygroscopic | |
Nickel(II) nitrate hexahydrate |
Safety and Handling
GHS H Statement
H272-H334-H350-H360-H372-H341-H318-H302-H332-H315-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
H272-H302+H332-H315-H317-H318-H334-H335-H341-H350i-H360D-H372
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NICKEL NITRATE
EINECSNumber : 236-068-5
RTECSNumber : QR7300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only