Learn More
Nickel(II) nitrate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals
$229.90 - $722.74
Chemical Identifiers
CAS | 13478-00-7 |
---|---|
Molecular Formula | H12N2NiO12 |
Molecular Weight (g/mol) | 290.79 |
MDL Number | MFCD00149805 |
InChI Key | AOPCKOPZYFFEDA-UHFFFAOYSA-N |
Synonym | nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 |
PubChem CID | 61630 |
SMILES | O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC223155000
|
Thermo Scientific Chemicals
223155000 |
500 g | Plastic bottle |
Each for $229.90
|
|
||||
AC223150025
|
Thermo Scientific Chemicals
223150025 |
2.5 kg | Plastic bottle |
Each for $722.74
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
For analysisChemical Identifiers
13478-00-7 | |
290.79 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
61630 |
H12N2NiO12 | |
MFCD00149805 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
137.0°C | |
13478-00-7,13138-45-9 | |
98.5 | |
Crystalline Powder | |
98.5% min. (Complexometry) | |
Ni(NO3)2·6H2O | |
15,6598 | |
Solubility in water: 940g/L (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
290.79 | |
290.8 | |
Analytical | |
2.05 |
56.7°C | |
Blue to Green | |
100.0 | |
500 g | |
H12N2NiO12 | |
MFCD00149805 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
nickel(2+);dinitrate;hexahydrate | |
61630 | |
99% | |
Plastic bottle | |
Nickel(II) nitrate hexahydrate, 99% |
Safety and Handling
GHS H Statement
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallow
GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Avoid release to the e
GHS Signal Word: Danger
RTECSNumber : QR7300000
RUO – Research Use Only