missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitric Acid, 10% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company 532016

 View more versions of this product

Catalog No. 5320-16


Explore available promotions

Description

Description

Specifications

Specifications

Nitric Acid
7697-37-2
500 mL
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
9.9 to 10.1%
Colorless to Light Yellow Liquid
Natural Poly Bottle
HNO3
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
O[N+]([O-])=O
63.01
CHEBI:48107

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
O[N+]([O-])=O
HNO3
MFCD00011349
944
nitric acid
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS Documents

Documents

Promotions

Promotions