missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitric Acid, 10% (v/v), Ricca Chemical
$111.59 - $1088.66
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
5320-16
|
Ricca Chemical Company
532016 |
500 mL | Natural Poly Bottle |
Each for $111.59
|
|
||||
5320-32
|
Ricca Chemical Company
532032 |
1 L | Natural Poly Bottle |
Each for $361.28
|
|
||||
53-205
|
Ricca Chemical Company
53205 |
20 L | Cubitainer |
Each for $1,088.66
|
|
||||
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Specifications
Colorless to Light Yellow Liquid | |
Natural Poly Bottle | |
HNO3 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
Nitric Acid |
7697-37-2 | |
500 mL | |
MFCD00011349 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
9.9 to 10.1% |
Safety and Handling
ShelfLife : 24 months