missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitric Acid, 10% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 532032
Specifications
Nitric Acid | |
7697-37-2 | |
1 L | |
MFCD00011349 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
9.9 to 10.1% |
Colorless to Light Yellow Liquid | |
Natural Poly Bottle | |
HNO3 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 |
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Safety and Handling
ShelfLife : 24 months