missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One Non-Vented Polystyrene Petri Dishes

Used for easy stacking

Supplier:  Greiner Bio-One 632181

 View more versions of this product

Catalog No. 07000341


Explore available promotions

Description

Description

  • For long incubation periods
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications

Specifications

80 mL
Polystyrene
Sterile
480
Standard
16 mm
Clear
Round
Standard
20
No
94 mm
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.