Honeywell™ 5500 Series Half-Mask Air Purifying Respirators
Incorporates the low-profile design of the North 7700 Series in an economical material. Cartridges and filters not included.
Supplier:  Honeywell™ 550030L
 View more versions of this product
Catalog No. 18-987C
Explore available promotions
Description
Description
  • Easy-to-clean facepiece conforms gently to wearer's face and is made from a soft elastomer that resists distortion
  • 'One-piece' cradle suspension system is easily removed for cleaning and provides a comfortable, secure fit with no slipping
  • Contoured sealing flange eliminates pressure points
  • Extended side flanges ensure secure seal even when wearer is talking
  • Exhalation valve assembly provides low breathing resistance and permits easy, positive pressure fit check without removing cover
  • Headband straps are adjustable and virtually impossible to lose or misassemble; headband yoke improves peripheral vision by allowing cartridges to be lower and farther back
  • Direct cartridge-to-facepiece seal eliminates risk of improper seal and reduced protection due to lost or worn sealing gaskets
  • Requires limited maintenance and available in three sizes that accept North cartridges and filters
Compliance
NIOSH approved under 42 CFR Part 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters.
Specifications
Specifications
MSHA
OSHA 29 CFR Part 1920.134
Gray
Half-Mask Respirator
North™ Cartridge, Filter and Accessories
NIOSH 42 CFR Part 84
Threaded
Thermoplastic Elastomer
Large
550030L
SDS
Documents
Documents
Accessories