missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ 5500 Series Half-Mask Air Purifying Respirators
Incorporates the low-profile design of the North 7700 Series in an economical material. Cartridges and filters not included.
$6.90 - $24.61
Specifications
Certifications/Compliance | MSHA |
---|---|
Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
Certifications (OSHA) | OSHA 29 CFR Part 1920.134 |
Connection Type | Threaded |
Color | Gray |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
18-987A
|
Honeywell™
550030S |
Small |
Each for $23.39
Case of 12 Each for $260.04
|
|
|||||
18-987B
|
Honeywell™
550030M |
Medium |
Each for $24.61
Case of 12 Each for $273.60
|
|
|||||
18-987C
|
Honeywell™
550030L |
Large |
Each for $6.90
Case of 12 Each for $82.80
|
|
|||||
Description
- Easy-to-clean facepiece conforms gently to wearer's face and is made from a soft elastomer that resists distortion
- 'One-piece' cradle suspension system is easily removed for cleaning and provides a comfortable, secure fit with no slipping
- Contoured sealing flange eliminates pressure points
- Extended side flanges ensure secure seal even when wearer is talking
- Exhalation valve assembly provides low breathing resistance and permits easy, positive pressure fit check without removing cover
- Headband straps are adjustable and virtually impossible to lose or misassemble; headband yoke improves peripheral vision by allowing cartridges to be lower and farther back
- Direct cartridge-to-facepiece seal eliminates risk of improper seal and reduced protection due to lost or worn sealing gaskets
- Requires limited maintenance and available in three sizes that accept North cartridges and filters
Compliance
NIOSH approved under 42 CFR Part 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters.
Specifications
MSHA | |
OSHA 29 CFR Part 1920.134 | |
Gray | |
Half-Mask Respirator |
NIOSH 42 CFR Part 84 | |
Threaded | |
Thermoplastic Elastomer | |
North™ Cartridge, Filter and Accessories |