Honeywell™ North™ 7700 Series Silicone Half-Mask Air-Purifying Respirator
Half-mask respirators with medical grade silicone sealing surface are flexible to provide a soft, secure, and comfortable fit. Low inhalation and exhalation resistance makes breathing easier to reduce worker fatigue.
Supplier:  Honeywell™ 770030S
 View more versions of this product
Catalog No. 17-620-1
Explore available promotions
Requires : Compatible with North cartridges, filters and accessories.
Description
Description
  • Soft, non-allergenic medical grade silicone provides protection, comfort and fit
  • Half-mask cradle suspension system allows facepiece to seal evenly on face without creating pressure points
  • Low profile provides a wide field of vision and does not interfere with protective eyewear
  • Minimal dead air space improves user comfort by limiting re-breathing of exhaled air
  • Direct cartridge-to-facepiece seal minimizes replacement parts and simplifies maintenance
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
NIOSH approved under 42 CFR 84 when used as designed with appropriate cartridges and filters.
Specifications
Specifications
NIOSH 42 CFR Part 84
Silicone
Small
770030S
Threaded
Half-Mask Respirator
North™ N-Series Cartridges, Filters and Accessories
Cradle
Videos
SDS
Documents
Documents
Accessories