missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ North™ 7700 Series Silicone Half-Mask Air-Purifying Respirator
Half-mask respirators with medical grade silicone sealing surface are flexible to provide a soft, secure, and comfortable fit. Low inhalation and exhalation resistance makes breathing easier to reduce worker fatigue.
$40.97 - $46.78
Specifications
Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
---|---|
Connection Type | Threaded |
Material | Silicone |
Product Type | Half-Mask Respirator |
For Use With (Equipment) | North™ N-Series Cartridges, Filters and Accessories |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-620-1
|
Honeywell™
770030S |
Small |
N/A
|
|
|||||
17-620-2
![]() |
Honeywell™
770030M |
Medium |
N/A
|
![]() |
|||||
17-620-3
|
Honeywell™
770030L |
Large |
N/A
|
|
|||||
Description
- Soft, non-allergenic medical grade silicone provides protection, comfort and fit
- Half-mask cradle suspension system allows facepiece to seal evenly on face without creating pressure points
- Low profile provides a wide field of vision and does not interfere with protective eyewear
- Minimal dead air space improves user comfort by limiting re-breathing of exhaled air
- Direct cartridge-to-facepiece seal minimizes replacement parts and simplifies maintenance
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
NIOSH approved under 42 CFR 84 when used as designed with appropriate cartridges and filters.
Specifications
NIOSH 42 CFR Part 84 | |
Silicone | |
North™ N-Series Cartridges, Filters and Accessories |
Threaded | |
Half-Mask Respirator | |
Cradle |