Honeywell™ North Adaptec™ Safety Glasses
Honeywell™ North Adaptec™ Safety Glasses deliver excellent protection, superior comfort, customized adjustability, premium optics and stylish look.
Supplier:  Honeywell™ T5900WTK
 View more versions of this product
Catalog No. 29-636-1705
Explore available promotions
Description
Description
  • Adjustable nose bridge centers eyes to reduce fatigue
  • Self-adjusting nose pads provde greater comfort, contact and stability
  • Vented cushioned frame allows optimum air circulation to reduce fogging
  • Temple hinge ratchets to seven positions — 30° of adjustment— to angle lens angle for optimal eye protection
  • Temple design adjusts tension for personal comfort
  • 99.9% UV protection
  • Replaceable lenses
  • Adjustable temples for comfort
  • Available in many lens tints, including SCT
Specifications
Specifications
Anti-fog, Anti-static, Scratch-resistant
Wide Size, Translucent-Black Frame, Clear Lens
Safety Glasses
Polycarbonate
Meets ANSI Z87.1-2010, CSA Z94.3
Clear
Translucent Black
Wide