Honeywell™ North™ 16 mil Butyl Chemical Resistant Gloves
Unsupported 16 mil thick butyl gloves with rolled beaded cuffs offer permeation resistance to gas and water vapor. Available in smooth finish or Grip-Saf™ palm for enhanced grip.
Supplier:  Honeywell™ B1617
 View more versions of this product
Catalog No. 19-025590
Explore available promotions
Description
Description
For use with ketones and esters, and applications such as aircraft maintenance, automotive, chemical handling and processing and hazmat work.
  • Made from butyl synthetic rubber
  • All gloves have rolled cuffs
  • Available with smooth or rough Grip Saf™ finish for wet applications
  • Unlined wtih sanitized interior
Specifications
Specifications
ASTM D3389-05, ASTM F790-97, OSHA 29 CFR, EN 374
Black
Yes
7
0.40 mm
Aircraft maintenance, chemical handling and processing, domestic preparedness, laboratory and hazmat
Smooth
B161/7
27.9 cm
EN 420
Specific
Butyl
16 mil
EN 388
Beaded/Rolled
Unlined
11 in.