Honeywell™ North™ 16 mil Butyl Chemical Resistant Gloves
Unsupported 16 mil thick butyl gloves with rolled beaded cuffs offer permeation resistance to gas and water vapor. Available in smooth finish or Grip-Saf™ palm for enhanced grip.
$99.34 - $100.88
Specifications
Certifications/Compliance ASTM D3389-05, ASTM F790-97, OSHA 29 CFR, EN 374
Certifications/Compliance (Europe) EN 420
Color Black
Hand Specific
Latex Free Yes
View More Specs
Products 2
Catalog Number Mfr. No. Size Grip Design Price Quantity  
Catalog Number Mfr. No. Size Grip Design Price Quantity  
19-025590
View Documents
Honeywell™
B1617
7 Smooth
Pair for $100.88
29-635-9733
View Documents
Honeywell™
B1619
9 Smooth
Pair for $99.34
Description
Description
For use with ketones and esters, and applications such as aircraft maintenance, automotive, chemical handling and processing and hazmat work.
  • Made from butyl synthetic rubber
  • All gloves have rolled cuffs
  • Available with smooth or rough Grip Saf™ finish for wet applications
  • Unlined wtih sanitized interior
Specifications
Specifications
ASTM D3389-05, ASTM F790-97, OSHA 29 CFR, EN 374
Black
Yes
16 mil
EN 388
Beaded/Rolled
11 in.
EN 420
Specific
Butyl
0.40 mm
Aircraft maintenance, chemical handling and processing, domestic preparedness, laboratory and hazmat
Unlined
27.9 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'