Honeywell™ North™ Primair™ 100 Series PA101 Loose-Fitting Facepiece Assembly
Loose-fitting assemblies are designed for pharmaceutical and other light industries.
Supplier:  Honeywell™ PA101S
 View more versions of this product
Catalog No. 19-134-240
Explore available promotions
Description
Description
  • Spun-bound polypropylene hood cover with Velcro™ strip and rust-resistant snaps attaches and removes easily from headgear
  • Triple adjustment headgear provides a secure and comfortable fit
  • Replaceable 3/16 in. (0.48 cm) foam sweatband with adhesive backing adheres and removes easily from headband
  • Lens made of 0.040 mil co-polyester
  • Polypropylene chin cover
  • Hoods with collar and full bib have an assigned protection factor (APF) of 1000
  • Manifold directs air over lens to reduce fogging
Note: At the current time, the color of product received may be yellow instead of the white material as seen in the photo above.  
Specifications
Specifications
NIOSH
Facepiece Assembly
Pharmaceutical and light duty applications
PA101L
Polypropylene
Large
Compact Air 200 Series PAPR and CF1000 Series