Honeywell™ North™ Primair™ 100 Series PA101 Loose-Fitting Facepiece Assembly
Loose-fitting assemblies are designed for pharmaceutical and other light industries.
$79.68 - $83.98
Specifications
Certifications/Compliance NIOSH
Material Polypropylene
Product Type Facepiece Assembly
For Use With (Application) Pharmaceutical and light duty applications
For Use With (Equipment) Compact Air 200 Series PAPR and CF1000 Series
View More Specs
Includes: Loose-fitting facepiece assemblies include adjustable headgear, facepiece, and a replacement sweatband. Hood assemblies include adjustable headgear, hood, sweatband, and peel-off cover lens.
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-134-241
View Documents
Honeywell™
PA101M
Medium
Each for $83.98
19-134-240
View Documents
Honeywell™
PA101S
Large
Each for $79.68
Description
Description
  • Spun-bound polypropylene hood cover with Velcro™ strip and rust-resistant snaps attaches and removes easily from headgear
  • Triple adjustment headgear provides a secure and comfortable fit
  • Replaceable 3/16 in. (0.48 cm) foam sweatband with adhesive backing adheres and removes easily from headband
  • Lens made of 0.040 mil co-polyester
  • Polypropylene chin cover
  • Hoods with collar and full bib have an assigned protection factor (APF) of 1000
  • Manifold directs air over lens to reduce fogging
Note: At the current time, the color of product received may be yellow instead of the white material as seen in the photo above.  
Specifications
Specifications
NIOSH
Facepiece Assembly
Compact Air 200 Series PAPR and CF1000 Series
Polypropylene
Pharmaceutical and light duty applications
SDS
missing translation for 'documents'
missing translation for 'documents'