Honeywell™ North™ Model 7190 N99 Air-Purifying Respirator
Economical and reusable
Supplier:  Honeywell™ 7190N99
 View more versions of this product
Catalog No. 27-057-23
Explore available promotions
Description
Description
  • Non-oil particulate Model 7190 N99 provides a cool, comfortable alternative to disposable respirators
  • Coaxial valving system provides separate intake and outflow channels
  • Larger filter surface area offers low breathing resistance
  • Low-profile design fits easily under a welding helmet with no obstruction of vision
  • Replaceable N99 filter provides protection against solid particulates and non–oil-based aerosols
Compliance
Respirator meets OSHA requirements and is approved to NIOSH 42CFR84 standards.
Specifications
Specifications
OSHA, NIOSH 42CFR84, ISO 9001
Single-use
Air Purifying Respirator
Provides protection against solid particulates and non-oil based aerosols
Each
Low profile
N99
One Size
(1 ) Respirator, (1) N99 Filter