Honeywell™ North™ Model 7190 N99 Air-Purifying Respirator
Economical and reusable
$330.80 - $330.80
Catalog Number | Mfr. No. | Product Type | Certifications/Compliance | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Certifications/Compliance | Price | Quantity | ||||
27-057-23
|
Honeywell™
7190N99 |
Air Purifying Respirator | OSHA, NIOSH 42CFR84, ISO 9001 |
Case of 12 Each for $330.80
|
|
||||
Description
- Non-oil particulate Model 7190 N99 provides a cool, comfortable alternative to disposable respirators
- Coaxial valving system provides separate intake and outflow channels
- Larger filter surface area offers low breathing resistance
- Low-profile design fits easily under a welding helmet with no obstruction of vision
- Replaceable N99 filter provides protection against solid particulates and non–oil-based aerosols
Compliance
Respirator meets OSHA requirements and is approved to NIOSH 42CFR84 standards.
Specifications
Low profile | |
N99 |