missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ SilverShield™ SSG Gloves
Unique five-layer fabric is resistant to over 280 different chemicals including alcohols, aliphatics, aromatice, chlorines, ketones and esters.
$361.11 - $1301.39
Specifications
Certifications/Compliance | OSHA 29 CFR 1915.157 |
---|---|
Certifications/Compliance (Europe) | EN 420, EN 374-3 |
Color | Silver |
Hand | Ambidextrous |
Latex Free | Yes |
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Size | Price | Quantity | ||||
18-999-431B
|
Honeywell™
SSG7 |
14.5 in. | 37 cm | 7 |
Case of 50 Pair for $457.05
|
|
||||
18-999-431C
|
Honeywell™
SSG8 |
14.5 in. | 37 cm | 8 |
Case of 50 Pair for $435.80
|
|
||||
18-999-431D
|
Honeywell™
SSG9 |
14.5 in. | 37 cm | 9 |
Case of 50 Pair for $361.11
|
|
||||
19-150-0892
|
Honeywell™
SSG2910 |
29 in. | 74 cm | 10 |
Case of 50 Each for $1,301.39
|
|
||||
Description
- Unique five-layer construction provides chemical protection
- Ideal for hospitals, agriculture, military and hazardous material handling
- Easily worn under heavy-duty outer gloves
- Use as a primary glove where there is minimal risk of tearing or puncturing
- Resistant to over 280 different chemicals, including alcohols, aliphatics, aromatics, chlorines, ketones and esters
- Does not contain latex or chemical accelerators
Specifications
OSHA 29 CFR 1915.157 | |
Silver | |
Yes | |
2.7 mil | |
EN 388 | |
Straight | |
Unlined |
EN 420, EN 374-3 | |
Ambidextrous | |
Polyethylene, EVA | |
0.06 mm | |
Bacteria, Fungi only | |
Smooth | |
SilverShield™ SSG |