missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell North™ North Zone™ Full Brim Hard Hat
Full brim hard hat with adjustable headband
Supplier: Honeywell North™ N20R030000
Description
- 3-level height adjustment (low, medium, high) for greater stability, balance and retention
- Removable
- Replaceable rear comfort cradle for improved fit and comfort
- Easy-grip ratchet for smooth and precise headband adjustment
- Even when wearing gloves
- Ergonomic headband for adjustment to 3 different sizes (small, medium/large and X-large)
- Moisture-wicking, breathable sweatband
- Easily removable, washable and replaceable
- Full brim and low-profile design for increased field of vision and user mobility
- Slots for cap-mounted earmuffs, face shields and other PPE accessories
Specifications
ANSI/ISEA Z89.1 2014 Type I, Class C, G, and E, CSA Z94.1 2015 Type I, Class C, G, and E | |
Full Brim Hard Hat | |
Not Ventilated | |
27.94 cm |
Orange | |
Universal | |
Full Brim | |
11 in. |