Honeywell North™ North Zone™ Short Brim Hard Hat
Short brim and low-profile design for increased field of vision and user mobility
Supplier:  Honeywell North™ N10100000
 View more versions of this product
Catalog No. 17378678
Explore available promotions
Description
Description
  • 3-level height adjustment (low, medium, high) for greater stability, balance and retention
  • Removable
    • Replaceable rear comfort cradle for improved fit and comfort
  • Easy-grip ratchet for smooth and precise headband adjustment
    • Even when wearing gloves
  • Ergonomic headband for adjustment to three sizes (small, medium/large, X-large)
  • Approved to be worn in reverse position
  • Moisture-wicking, breathable sweatband
    • Easily removable, washable and replaceable
Specifications
Specifications
ANSI/ISEA Z89.1 2014: Type I, Class C, G and E, CSA Z94.1 2015: Type I, Class C, G and E
Low-profile Hard Hat
Quick fit
Short Brim
8.6 in.
Tan
Universal
Not Ventilated
21.844 cm